Learn More
Sampling Systems™ GMP Pallet Knife
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
190 mm
240 mm
325 mm
380 mm
440 mm
490 mm
Unit Size:
Each
Description
- Strong and flexible
- Range of different lengths
Specifications
Specifications
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Shape | Curved |
| Disposable | Reusable |
| Product Type | Pallet Knife |
| Material | Stainless Steel |
| Length (Metric) | 240 mm |