Learn More
Thermo Scientific™ GNE7915, 95%
Brand: Thermo Scientific™ 468021000
450.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
GNE7915 | |
[4-[[4-(Ethylamino)-5-(trifluoromethyl)-2-pyrimidinyl]amino]-2-fluoro-5-methoxyphenyl]-4-morpholinyl-methanone | |
C19 H21 F4 N5 O3 | |
Solid | |
95% |
1351761-44-8 | |
100 mg | |
443.40 | |
Reagent |