Learn More
Goat anti-Mouse IgG1 Secondary Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69522
This item is not returnable.
View return policy
Description
IgG1 Polyclonal antibody specifically detects IgG1 in Mouse samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)Specifications
IgG1 | |
Polyclonal | |
FITC | |
Lyophilized from PBS (pH 7.2) | |
Goat | |
Ion exchange chromatography | |
RUO | |
Secondary | |
Mouse | |
Lyophilized |
ELISA, Immunofluorescence, Immunohistochemistry (Frozen) | |
LYOPH | |
ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen | |
IgG1, Igh-4, RGD1359539, VH7183 | |
Pools of highly purified homogenous IgG1 isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
1 mL | |
Immunology | |
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
Store at 4°C in the dark. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu