missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit Ig, Polyclonal, HRP, Secondary Antibody, BD
Brand: BD Biosciences 554021
This item is not returnable.
View return policy
Description
X
Host Species: Goat
Species Reactivity: Rabbit
Flow Cytometry
Specifications
Goat anti-Rabbit Ig | |
POLYCLONAL | |
1 mL | |
Immunology | |
Rabbit |
Polyclonal | |
HRP | |
RUO | |
Secondary | |
Ig |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction