Learn More
Goat anti-Rat IgE Fc Secondary Antibody, TRITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69135
Product Code. 18643307
  • 626.00 EUR / 1mL
Explore more special offers
Description
Description
IgE Fc Polyclonal antibody specifically detects IgE Fc in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
Specifications
IgE Fc
Polyclonal
TRITC
Lyophilized from PBS (pH 7.2)
Pools of purified homogenous IgE isolated from rat serum. Freund's complete adjuvant is used in the first step of the immunization procedure
1 mL
Secondary
Rat
Lyophilized
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
LYOPH
ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Goat
Ion exchange chromatography
RUO
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C in the dark.