Learn More
Goat anti-Rat IgE Fc Secondary Antibody, TRITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69135
This item is not returnable.
View return policy
Description
IgE Fc Polyclonal antibody specifically detects IgE Fc in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)Specifications
IgE Fc | |
Polyclonal | |
TRITC | |
Lyophilized from PBS (pH 7.2) | |
Pools of purified homogenous IgE isolated from rat serum. Freund's complete adjuvant is used in the first step of the immunization procedure | |
1 mL | |
Secondary | |
Rat | |
Lyophilized |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
LYOPH | |
ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
Goat | |
Ion exchange chromatography | |
RUO | |
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
Store at 4°C in the dark. |