missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG2a Secondary Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69582
This item is not returnable.
View return policy
Description
IgG2a Polyclonal antibody specifically detects IgG2a in Rat samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)Specifications
IgG2a | |
Polyclonal | |
HRP | |
Lyophilized from PBS (pH 7.2) | |
Pools of purified homogenous IgG2a isolated from pooled rat serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
1 mL | |
Secondary | |
Rat | |
Lyophilized |
ELISA, Immunofluorescence, Immunohistochemistry (Frozen) | |
LYOPH | |
ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen | |
Goat | |
Ion exchange chromatography | |
RUO | |
Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction