Learn More
3M™ GoggleGear™ 6000 Safety Goggles
GoggleGear™ Safety Goggles 6000 Series features a large profile with indirect ventilation for air circulation with Scotchgard™ Anti-Fog and Anti-Scratch Coating.
Brand:  3M™ 7100216191
237.81 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17659725
  • 347.00 EUR / Pack of 10
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Description
  • For safety goggles with indirect ventilation and a 3M™ Scotchgard™ Anti-Fog and Anti-Scratch coating, use 3M™ GoggleGear™ Safety Goggles 6000 Series.
  • Our large profile safety goggles feature indirect ventilation to allow air to circulate and help reduce fogging.
  • The polycarbonate lens features a 3M™ Scotchgard™ Anti-Fog and Anti-Scratch coating for superior anti-fog protection and increased durability that resists fogging for longer than traditional anti-fog coatings, even after up to 25 washings with water*.
  • Available in a range of shroud colours and two lens options.
  • *3M internal testing per EN168 test method when compared with traditional anti-fog coatings.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Anti-fog
Safety Goggles
Polypropylene
Polycarbonate
UV400
None
Blue
Reusable
Polypropylene
Large Profile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ GoggleGear™ 6000 Safety Goggles > Blue Shroud