Learn More
Gosselin™ T-shaped Polystyrene Spreader
Click to view available options
Quantity:
5 Boxes x 18 zip bags of 50
Unit Size:
Pack of 4500
Description
- Polystyrene (PS)
- Packed in resealable bag
- Curved end or 'T' end
- Sterile
Specifications
Specifications
| Disposable | Single-use |
| Color | Green |
| Length (Metric) | 148 mm |
| Quantity | 5 Boxes x 18 zip bags of 50 |
| Shape | T-Shape |
| Sterility | Sterile |
| For Use With (Application) | For Dairy, Environmental, Industrial, Microbiological Uses |