missing translation for 'onlineSavingsMsg'
Learn More

Gossypol-Acetic Acid, MP Biomedicals™

Product Code. 11471122 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
This item is not returnable. View return policy

Product Code. 11471122

Brand: MP Biomedicals 0219521180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 12542-36-8
Molecular Formula C32H34O10
Molecular Weight (g/mol) 578.614
InChI Key NIOHNDKHQHVLKA-UHFFFAOYSA-N
Synonym gossypol-acetic acid, gossypol acetate, gossypol acetic acid, acetate gossypol, --gossypol acetic acid, s-gossypol acetic acid, at-101 acetic acid, gossypol-acetic acid complex
PubChem CID 227456
IUPAC Name acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
SMILES CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O

Specifications

Quantity 100 mg
Formula Weight 578.61
Physical Form Solid
Chemical Name or Material Gossypol-Acetic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.