missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPAT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA557844
This item is not returnable.
View return policy
Description
Immunogen sequence: HQKGVFLSQL LGEFSWLTEE ILLRGFDVGF SGQLRSLLQH SLSLLRAHVA LLRIRQGDLL VVPQPGPGLT HLA Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
GPAT2 gene ontology annotations related to this gene include cellular lipid metabolic process.Specifications
GPAT2 | |
Polyclonal | |
Unconjugated | |
GPAT2 | |
A530057A03Rik; cancer/testis antigen 123; CT123; Glycerol-3-phosphate acyltransferase 2, mitochondrial; glycerol-3-phosphate acyltransferase-like 1; Gm116; GPAT2; GPAT-2; RGD1304904; xGPAT1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150763 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NUI2 | |
GPAT2 | |
Recombinant protein corresponding to Human GPAT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction