missing translation for 'onlineSavingsMsg'
Learn More

GRA Ex-25, MedChemExpress

Product Code. 16473997
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473997

Brand: Medchem Express HY50675

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GRA Ex-25 is an inhibitor of glucagon receptor, with IC50 of 56 and 55 nM for rat and human glucagon receptors, respectively.

GRA Ex-25 is an inhibitor of glucagon receptor, with IC50 of 56 and 55 nM for rat and human glucagon receptors, respectively.

Specifications

Chemical Name or Material GRA Ex-25
CAS 307983-31-9
Color White
Purity Grade Notes Research
Molecular Formula C29H36F3N3O5
Quantity 50 mg
Solubility Information DMSO : ≥ 32 mg/mL (56.78 mM)
SMILES O=C(O)CCNC(C1=CC=C(CN([C@H]2CC[C@H](C(C)(C)C)CC2)C(NC3=CC=C(OC(F)(F)F)C=C3)=O)C=C1)=O
Molecular Weight (g/mol) 563.61
Formula Weight 563.61
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.