Learn More
asecos™ Granulate Universal Fine Grain
Product Code. 17826376 Shop All asecos Products
Click to view available options
Weight (Metric):
20 kg
60 kg
Unit Size:
Each
Pack of 3
Product Code. 17826376
Brand: asecos™ 18453
This item requires a price update prior to being ordered.
Request a Price for this item here.
Granulate Universal Fine Grain is ideal for setting oils, alkalis, lipids, fuels, lacquers etc.
  • 0.5-1 mm grain size
  • 20 kg bag binds up to 18 L heating oil
  • Penetrates small depressions and cracks
  • Best suited for large contact with the surface and fast absorption (e.g. traffic surfaces)
  • Optimum slip resistance even when completely saturated
  • Available in one or three 20 kg bags.
Specifications
Material Loose Particulate
Type Absorbent Granules
Weight (English) 132.28 lb.
Weight (Metric) 60 kg