Learn More
Thermo Scientific™ Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Improve your AAS graphite furnace performance using Thermo Scientific™ application-specific graphite cuvettes.
1445.00 EUR
Specifications
Type | Graphite Furnace Cuvette |
---|---|
For Use With (Equipment) | GFS33 or GFS35 Furnaces |
Description
Discover different types of graphite furnace cuvettes suitable for any analytical application using the Thermo Scientific™ GFS33/GFS35/GFS35Z Graphite Furnace and Autosampler modules
Improve instrument performance with the right selection of graphite cuvettesWhile ridged cuvettes are supplied as standard with Thermo Scientific GFS33/GFS35/GFS35Z Graphite Furnaces, additional application-specific cuvettes improve performance for more complex applications:- Extended Lifetime Cuvettes (ELCs) can be used longer and provide reproducible performance over several thousand firings
- Ridged Cuvettes (in pyrolytically-coated and uncoated forms) help maintain difficult samples at the optimum temperature for efficient atomization
- Omega Cuvettes with integral platforms provide the most interference-free conditions (often in combination with matrix modification) for samples with complex matrices
Specifications
Graphite Furnace Cuvette | |
GFS33 or GFS35 Furnaces |