missing translation for 'onlineSavingsMsg'
Learn More

Grifolic acid, Tocris Bioscience™

Product Code. 18035442
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective partial FFA4 (GPR120) agonist

Selective partial FFA4 (GPR120) agonist. Induces ERK and [Ca2+]i responses in cells expressing GPR120, but not those expressing GPR40.

Specifications

CAS 80557-12-6
Molecular Formula C23H32O4
Quantity 1 mg
Synonym grifolic acid, ilicicolinic acid b, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyldodeca-2,6,10-trienyl benzoic acid, d0e9ay, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyl-2,6,10-dodecat rien-1-yl benzoic acid, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyl-2,6,10-dodecatrien-1-yl-benzoic acid, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyldodeca-2,6,10-trien-1-yl benzoic acid, benzoic acid, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyl-2,6,10-dodecatrienyl
InChI Key QPIZDZGIXDKCRC-JTCWOHKRSA-N
SMILES CC1=CC(=C(C(=C1C(=O)O)O)CC=C(C)CCC=C(C)CCC=C(C)C)O
IUPAC Name 2,4-dihydroxy-6-methyl-3-[(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl]benzoic acid
Molecular Weight (g/mol) 372.505
PubChem CID 9976563
Formula Weight 372.5
Percent Purity >98%
Chemical Name or Material Grifolic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.