Learn More
Microspec™ Grip Seal Bags
Click to view available options
Dimensions (L x W):
2 x 9 in.
3 x 7.5 in.
6 x 9 in.
7.5 x 7.5 in.
Unit Size:
Pack of 1000
Description
- Grip Seal Bag
- Sterile
- Pack of 1000
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Disposable | Single-use |
| For Use With (Application) | Sample Transport |
| Sterility | Sterile |
| Film Description | LDPE |
| No. per Case | 1000 |
| No. per Pack | 100 |
| Sterilization Method | EtO |
| Dimensions (L x W) | 6 x 9 in. |