missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSDMD, Mouse anti-Human, Clone: 3F12-1B2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00079792-M01.100ug
65.02 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 0.00010kg
Description
Sequence: MGSAFERVVRRVVQELDHGGEFIPVTSLQSSTGFQPYCLVVRKPSSSWFWKPRYKCVNLSIKDILEPDAAEPDVQRGRSFHFYDAMDGQIQGSVELAAPGQAKIAGGAAVSDSSSTSMNVYSLSVDPNTWQTLLHERHLRQPEHKVLQQLRSRGDNVYVVTEVLQTQKEVEVTRTHKREGSGRFSLPGATCLQGEGQGHLSQKKTVTIPSGSTLAFRVAQLVIDSDLDVLLFPDKKQRTFQPPATGHKRSTSEGAWPQLPSGLSMMRCLHNFLTDGVPAEGAFTEDFQGLRAEVETISKELELLDRELCQLLLEGLEGVLRDQLALRALEEALEQGQSLGPVEPLDGPAGAVLECLVLSSGMLVPELAIPVVYLLGALTMLSETQHKLLAEALESQTLLGPLELVGSLLEQSAPWQERSTMSLPPGLLGNSWGEGAPAWVLLDECGLELGEDTPHVCWEPQAQGRMCALYASLALLSGLSQEPHSpecifications
GSDMD | |
Monoclonal | |
Unconjugated | |
gasdermin D | |
DF5L/FKSG10/FLJ12150/GSDMDC1 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3F12-1B2 | |
In 1x PBS, pH 7.4 | |
BC008904 | |
GSDMD | |
GSDMD (AAH08904, 1 a.a. ∼ 484 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
79792 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |