missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSE1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
360.68 EUR - 575.58 EUR
Specifications
Antigen | GSE1 |
---|---|
Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18320847
|
Bio-Techne
NBP3-17501-25UL |
25 μg |
360.68 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18331335
|
Bio-Techne
NBP3-17501-100UL |
100 μg |
575.58 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSE1 Polyclonal antibody specifically detects GSE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
GSE1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
KIAA0182 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: VSLSEPATQQASLDVEKPVGVAASLSDIPKAAEPGKLEQVRPQELSRVQELAPASGEKARLSEAPGGKKSLSMLHYIRG | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
23199 | |
IgG | |
Affinity purified |