Learn More
Tocris Bioscience™ GSK 1562590 hydrochloride
High affinity, selective urotensin II (UT) receptor antagonist
Brand:  Tocris Bioscience™ 5110/50
Encompass
Product Code. 15766476
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
GSK 1562590 hydrochloride is a high affinity and selective urotensin II (UT) receptor antagonist (pKi values are 9.14, 9.28, 9.34, 9.64 and 9.66 at monkey, human, mouse, cat and rat recombinant receptors respectively). Exhibits selectivity for UT receptors over a range of GPCRs, ion channels, enzymes and neurotransmitter transporters. Supresses human urotensin-II (hU-II)-induced contraction of isolated rat aorta in vitro and ex vivo. Inhibits the hU-II-induced increase in mean blood pressure in vivo. Orally active.
Specifications
Specifications
>98%