missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine-5'-monophosphate, Free acid, ≥98%, MP Biomedicals™
30.55 EUR - 74.70 EUR
Chemical Identifiers
CAS | 85-32-5 |
---|---|
Molecular Formula | C10H14N5O8P |
Molecular Weight (g/mol) | 363.223 |
InChI Key | RQFCJASXJCIDSX-UUOKFMHZSA-N |
Synonym | 5'-guanylic acid, guanosine monophosphate, guanylic acid, guanosine-5'-monophosphate, guanosine 5'-monophosphate, 5'-gmp, guanosine 5'-phosphate, guanylate, guanidine monophosphate, guanosine-phosphate |
PubChem CID | 6804 |
ChEBI | CHEBI:17345 |
IUPAC Name | [(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
SMILES | C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)NC(=NC2=O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394359
|
MP Biomedicals
0210190880 |
100 mg |
30.55 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304369
|
MP Biomedicals
0210190890 |
500 mg |
74.70 EUR
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314369
|
MP Biomedicals
0210190891 |
1 g |
70.85 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
85-32-5 | |
363.223 | |
5'-guanylic acid, guanosine monophosphate, guanylic acid, guanosine-5'-monophosphate, guanosine 5'-monophosphate, 5'-gmp, guanosine 5'-phosphate, guanylate, guanidine monophosphate, guanosine-phosphate | |
CHEBI:17345 | |
C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)NC(=NC2=O)N |
C10H14N5O8P | |
RQFCJASXJCIDSX-UUOKFMHZSA-N | |
6804 | |
[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
Specifications
85-32-5 | |
≥98% | |
5'-guanylic acid, guanosine monophosphate, guanylic acid, guanosine-5'-monophosphate, guanosine 5'-monophosphate, 5'-gmp, guanosine 5'-phosphate, guanylate, guanidine monophosphate, guanosine-phosphate | |
C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)NC(=NC2=O)N | |
363.223 | |
CHEBI:17345 | |
Powder |
White | |
C10H14N5O8P | |
RQFCJASXJCIDSX-UUOKFMHZSA-N | |
[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate | |
6804 | |
363.2 | |
Guanosine-5'-monophosphate, FTIR |
Safety and Handling
missing translation for 'einecsNumber' : 201-598-8