missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-Triphosphate Trisodium Salt 98-100%, MP Biomedicals™

Product Code. 11403751
Click to view available options
Quantity:
10 mg
25 mg
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 36051-31-7
Molecular Formula C10H16MgN5O14P3
Molecular Weight (g/mol) 547.49
MDL Number MFCD00077781,MFCD00077781
InChI Key NMQPZZLDIUETGE-GWTDSMLYSA-N
Synonym 5'-gtp trisodium salt, 5 acute-gtp trisodium salt
PubChem CID 92044363
SMILES [Mg].NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Color White
Assay Percent Range 98 to 100%
Quantity 10 mg
IUPAC Name ({[({[(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid magnesium
Formula Weight 588.2 (Anhydrous)
Physical Form Powder
Chemical Name or Material Guanosine-5'-Triphosphate Trisodium Salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'