Learn More
Gilac™ HACCP Flat Tray
Product Code. p-200009550
Click to view available options
Capacity (Metric):
3 L
5 L
8 L
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Product Code. 17928961
Brand: Gilac™ G155221
Only null left
Add to basket
Add to basket
HACCP Flat Tray can be used in a wide range of temperatures and its dimensions are suitable for refrigerators.
  • Flat trays.
  • Large and customisable range.
  • 5 colors.
  • 5 volumes.
  • Can be used in a wide range of temperatures.
  • Dimensions suitable for refrigerators.
  • Can be used as a:
    • defrosting tray thanks to its wide dividing grid that separates the ice water from the product being defrosted.
    • preparation container for products that drip.
    • ingredients tray.
    • container for preserving comparison samples.
    • Fermenting and decanting containers.
  • Nesting and stacking with lid.
  • Traceability management thanks (IML) injection molding.
Specifications
Capacity (Metric) 3 L
Color White
Height (Metric) 70 mm
Length (Metric) 347 mm
Material HDPE
Type Dispensing Tray
Width (Metric) 230 mm
Length (English) 13.7 in.
Height (English) 2.8 in.
Width (English) 9.1 in.
Capacity (English) 0.8 gal.