Learn More
Hamilton™ Large Hub Removable Needles (250μL - 10mL Syringes)
Product Code. p-4121860
Click to view available options
Needle Gauge:
22 G
22s G
26 G
26s G
Needle Point Style:
Blunt
Cone
Sharp, Bevel, Curved
Diameter (English) Inner:
0.0051 in.
0.0059 in.
0.01 in.
0.016 in.
Diameter (English) Outer:
0.018 in.
0.028 in.
Unit Size:
Pack of 6
Product Code. 10103253
Brand: Hamilton™ 778404
Large Hub Removable (RN) needles can be used with removable needle (RN)-style syringes that are 250μL and larger. Standard needle length is 2 in. (51mm). Custom gauges, lengths, and point styles are available.
Large Hub Removable Needle Compatible Syringe Terminations
  • Removable Needle (RN)- The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
  • Needle Point Styles
    • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 26-22
    • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 26-22.
    • Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-22
    Six needles per pack.
    Specifications
    Autoclavable Autoclavable
    Material Stainless Steel (Hub)
    Needle Gauge 26 G
    Diameter (English) Inner 0.01 in.
    Diameter (English) Outer 0.018 in.
    Diameter (Metric) Inner 0.26 mm
    Diameter (Metric) Outer 0.46 mm
    Length (English) Needle 2 in.
    Product Type Replacement Syringe Needle
    Needle Point Style Cone
    For Use With (Equipment) Removable Syringes (250μL and larger)