Learn More
Hamilton™ Metal Hub Blunt Point Needles (Luer Lock)
Click to view available options
Needle Gauge:
14 G
17 G
18 G
19 G
20 G
21 G
22 G
22s G
23 G
24 G
25 G
26 G
26s G
33 G
Unit Size:
Pack of 6
Description
Can be used with TLL syringes and LT or TLL connectors.
- Made of 304 Stainless Steel
- Standard needle length is 51mm (2 in.)
- Available in convenient six packs
- Custom needle lengths and point styles are available
- Needles are available from 10 to 33 ga
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Nickel-plated Brass (Hub), Stainless Steel |
| Needle Gauge | 22s G |
| Diameter (English) Inner | 0.005 in. |
| Diameter (English) Outer | 0.028 in. |
| Diameter (Metric) Inner | 0.15 mm |
| Diameter (Metric) Outer | 0.72 mm |
| Length (English) Needle | 2 in. |
| Product Type | Needle |
| Needle Point Style | Blunt |
| Show More |