Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
For use with MICROLAB 600 Diluter/Dispenser
147.00 EUR - 378.00 EUR
Product Code | Brand | Model | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Volume (Metric) | Price | Quantity & Availability | ||||
15142644
|
Hamilton™
59000-05 |
1701 BFP | 10 μL |
225.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
15122644
|
Hamilton™
59000-15 |
1705 BFP | 50 μL |
147.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
12638473
|
Hamilton™
59000-20 |
1710 BFP | 100 μL |
147.00 EUR
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
||||
15146862
|
Hamilton™
59000-10 |
1702 BFP | 25 μL |
147.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
15132644
|
Hamilton™
59000-10 |
1702 BFP | 25 μL |
147.00 EUR
Each |
Estimated Shipment: 08-11-2024
Log in to see stock. |
||||
12396239
|
Hamilton™
59000-25 |
1725 BFP | 250 μL |
147.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
13478496
|
Hamilton™
59000-30 |
1750 BFP | 500 μL |
150.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
12528126
|
Hamilton™
59000-35 |
1001 BFP | 1 mL |
160.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
13346319
|
Hamilton™
59000-40 |
1002 BFP | 2.5 mL |
177.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
11684133
|
Hamilton™
59000-50 |
1010 BFP | 10 mL |
199.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
15182644
|
Hamilton™
59000-60 |
Bubble free prime | 50 mL |
373.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
15192644
|
Hamilton™
59000-55 |
Bubble free prime | 25 mL |
378.00 EUR
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
12316249
|
Hamilton™
59000-45 |
1005 BFP | 5 mL |
227.00 EUR
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
||||
Description
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Specifications
Microlab™ 600 dispensers |