Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Black tips for accurate sample delivery
Brand: Axygen™ TT-20-CBK-HTR
This item is not returnable.
View return policy
Description
- Polypropylene
- Volume ranging from 20 to 1000μL
- Available with or without aerosol-resistant filters
- Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems
Certifications
Certified DNAse-, RNAse- and Pyrogen-safe
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
20 μL | |
Non-sterile | |
Autoclavable | |
For accurate sample delivery | |
Automation, Round | |
Polypropylene |
Non-filtered | |
Black | |
Non-ANSI | |
Hanging Tip Racked | |
Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head |