Learn More
Hanna™ Instruments pH Buffer Solutions
Product Code. 11504287
Click to view available options
Quantity:
230 mL
Unit Size:
Each
Product Code. 11504287
Brand: Hanna Instruments HI7004M
Used for calibration
  • Available in different pH ranges and quantity
  • Calibrated according to NIST standards
  • With/without certificates of analysis
Specifications
Quantity 230 mL