Learn More
Brady™ Hard Aluminum Safety Signs
11.22 EUR - 27.01 EUR
Specifications
Material | Aluminum |
---|---|
Quantity | 1 Ea. |
Description | Hard Aluminum |
For Use With (Application) | Used for indoor and outdoor safety signs |
Shape | Triangular |
Product Code | Brand | Displayed Text | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Width (Metric) | Price | Quantity & Availability | ||||
11780853
|
Brady™
807704 |
Corrosive | 100 mm | N/A | |||||
11780863
|
Brady™
807747 |
DANGER of Suffocation | 200 mm | N/A | |||||
11720883
|
Brady™
807764 |
Harmful or Irritating Material | 100 mm | N/A | |||||
11806361
|
Brady™
807767 |
Harmful or Irritating Material | 200 mm | N/A | |||||
11750883
|
Brady™
807770 |
Harmful or Irritating Material | 315 mm | N/A | |||||
11790883
|
Brady™
807774 |
Oxidizer | 100 mm | N/A | |||||
11720893
|
Brady™
807780 |
Oxidizer | 315 mm | N/A | |||||
Specifications
Aluminum | |
Hard Aluminum | |
Triangular |
1 Ea. | |
Used for indoor and outdoor safety signs | |
Safety Sign |