Learn More
3M™ Hard Hat with Plastic Sweatband, G2000CUV-VI
Product Code. 15453355
Click to view available options
:
Color: White
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 15453355
Brand: 3M™ 7000039611
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
3M™ Hard Hat G2000CUV-VI features a ventilation inlet on the top and a ventilated plastic headband to provide comfort.
  • Lightweight white hard hat with pinlock suspension
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV radiation and it's time to replace it
  • Extended edge on the back of the hard hat provides extra protection for the sensitive neck area
  • Flat surface for printing a brand name or logo
  • Easy to replace sweatband for comfort and hygiene
  • Ventilated headband for improved comfort
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN397
Color White
Material UV-Stabilized ABS
Size 54 to 62 cm
Weight (Metric) 340 g
Ventilated Ventilated