Learn More
Brady™ Harsh Environment Multipurpose Die-cut Opaque Polyester Labels for M6/M7 Printers
Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
26.60 EUR - 484.00 EUR
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | Halogen-free (DIN VDE 0472 Part 815) |
Adhesive Type | Permanent Acrylic |
Coating | None |
Color | White |
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Shape | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Shape | Price | Quantity & Availability | ||
17838774
|
Brady™
174190 |
0.187 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 4.75 mm | Rectangle |
78.55 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17858474
|
Brady™
174189 |
0.25 x 0.25 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Square |
42.85 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17818264
|
Brady™
174173 |
0.25 x 0.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Rectangle |
69.60 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17888124
|
Brady™
174186 |
0.25 x 0.9 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Rectangle |
74.15 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17898824
|
Brady™
174244 |
0.25 x 2 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Rectangle |
28.75 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17838264
|
Brady™
174246 |
0.25 x 1.25 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Rectangle |
85.10 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17888274
|
Brady™
174250 |
0.25 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Rectangle |
96.80 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17818414
|
Brady™
174334 |
0.2 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 5.08 mm | Rectangle |
60.65 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17858264
|
Brady™
174372 |
0.2 x 0.65 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 5.08 mm | Rectangle |
67.35 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17868744
|
Brady™
173434 |
0.25 x 0.75 in. | M710, BMP71 | 6.35 mm | Rectangle |
64.45 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17839064
|
Brady™
173423 |
0.25 x 0.25 in. | M710, BMP71 | 6.35 mm | Square |
39.70 EUR
Pack of 750 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||
17869174
|
Brady™
173371 |
0.25 x 2 in. | M710, BMP71 | 6.35 mm | Rectangle |
26.60 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17869144
|
Brady™
174590 |
0.25 x 1.25 in. | M710, BMP71 | 6.35 mm | Rectangle |
78.80 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17859224
|
Brady™
173366 |
0.25 x 1.5 in. | M710, BMP71 | 6.35 mm | Rectangle |
89.60 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17828294
|
Brady™
173318 |
0.2 x 0.5 in. | M710, BMP71 | 5.08 mm | Rectangle |
56.15 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17808134
|
Brady™
173317 |
0.275 x 0.5 in. | M710, BMP71 | 6.99 mm | Rectangle |
59.10 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17828184
|
Brady™
173304 |
0.2 x 0.65 in. | M710, BMP71 | 5.08 mm | Rectangle |
62.35 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17858304
|
Brady™
174337 |
0.275 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.99 mm | Rectangle |
63.85 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17878404
|
Brady™
174287 |
0.375 x 0.375 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 9.53 mm | Square |
44.95 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17888774
|
Brady™
173333 |
0.375 x 0.375 in. | M710, BMP71 | 9.53 mm | Square |
41.60 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17848454
|
Brady™
174197 |
0.375 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 9.53 mm | Rectangle |
66.65 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17868804
|
Brady™
174431 |
0.375 x 1 in. | M610, M710, M611, BMP61, BMP71 | 9.53 mm | Rectangle |
420.00 EUR
Pack of 5000 |
In Stock
Log in to see stock. |
||
17848464
|
Brady™
174249 |
0.125 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 3.18 mm | Rectangle |
87.15 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17808194
|
Brady™
174326 |
0.4 x 0.4 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 10.16 mm | Square |
46.90 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17818324
|
Brady™
174104 |
0.6 x 1.9 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 15.24 mm | Rectangle |
60.65 EUR
Pack of 250 |
In Stock
Log in to see stock. |
||
17898344
|
Brady™
173321 |
0.4 x 0.4 in. | M710, BMP71 | 10.16 mm | Square |
43.50 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17878414
|
Brady™
173313 |
0.5 x 0.5 in. | M710, BMP71 | 12.7 mm | Square |
54.05 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17899054
|
Brady™
173363 |
0.5 x 1.5 in. | M710, BMP71 | 12.7 mm | Rectangle |
68.90 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17888244
|
Brady™
174345 |
0.5 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Square |
58.35 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17808144
|
Brady™
174256 |
0.5 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Rectangle |
74.35 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17848164
|
Brady™
174207 |
0.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Rectangle |
66.65 EUR
Pack of 500 |
In Stock
Log in to see stock. |
||
17888204
|
Brady™
174493 |
0.5 x 0.5 in. | M610, M710, M611, BMP61, BMP71 | 12.7 mm | Square |
394.00 EUR
Pack of 5000 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
||
17848114
|
Brady™
174468 |
0.5 x 1.5 in. | M610, M710, M611, BMP61, BMP71 | 12.7 mm | Rectangle |
484.00 EUR
Pack of 5000 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||
17848764
|
Brady™
173445 |
0.5 x 1 in. | M610, M611, M710, BMP61, BMP71 | 12.7 mm | Rectangle |
432.00 EUR
Pack of 5000 |
In Stock
Log in to see stock. |
||
17818154
|
Brady™
174193 |
0.275 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.99 mm | Rectangle |
80.95 EUR
Pack of 750 |
In Stock
Log in to see stock. |
||
17808414
|
Brady™
174099 |
0.75 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm | Rectangle |
59.60 EUR
Pack of 250 |
In Stock
Log in to see stock. |
||
17809144
|
Brady™
173367 |
0.125 x 1.5 in. | M710, BMP71 | 3.18 mm | Rectangle |
80.70 EUR
Pack of 750 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||
17814758
|
Brady™
173346 |
1 x 1.5 in. | M710, BMP71 | 25.4 mm | Rectangle |
44.45 EUR
1 roll |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17879094
|
Brady™
173374 |
1 x 3 in. | M710, BMP71 | 25.4 mm | Rectangle |
38.25 EUR
Pack of 100 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||
17879044
|
Brady™
173373 |
1 x 4 in. | M710, BMP71 | 25.4 mm | Rectangle |
45.75 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17801429
|
Brady™
174274 |
1 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
48.10 EUR
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17878114
|
Brady™
174282 |
1.5 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 38.1 mm | Square |
75.85 EUR
Pack of 250 |
In Stock
Log in to see stock. |
||
17849044
|
Brady™
173369 |
1.25 x 2.75 in. | M710, BMP71 | 31.75 mm | Rectangle |
49.75 EUR
Pack of 100 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||
17819144
|
Brady™
174453 |
1 x 1 in. | M610, M710, M611, BMP61, BMP71 | 25.4 mm | Square |
306.00 EUR
Pack of 2500 |
In Stock
Log in to see stock. |
||
17879124
|
Brady™
173448 |
1 x 2 in. | M610, M611, M710, BMP61, BMP71 | 25.4 mm | Rectangle |
245.00 EUR
Pack of 1000 |
In Stock
Log in to see stock. |
||
17891409
|
Brady™
174220 |
1 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Square |
42.60 EUR
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17888334
|
Brady™
174240 |
1 x 3 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
41.30 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17828444
|
Brady™
174242 |
1 x 4 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
49.35 EUR
Pack of 100 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||
17838434
|
Brady™
174292 |
1.9 x 3 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 48.26 mm | Rectangle |
53.70 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17818284
|
Brady™
174299 |
1.9 x 4 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 48.26 mm | Rectangle |
66.95 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17878894
|
Brady™
173338 |
1.5 x 1.5 in. | M710, BMP71 | 38.1 mm | Square |
70.25 EUR
Pack of 250 |
In Stock
Log in to see stock. |
||
17848894
|
Brady™
173329 |
1.9 x 3 in. | M710, BMP71 | 48.26 mm | Rectangle |
49.75 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17808954
|
Brady™
173326 |
1.9 x 4 in. | M710, BMP71 | 48.26 mm | Rectangle |
62.05 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17898174
|
Brady™
174228 |
2 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 50.8 mm | Rectangle |
37.75 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17849024
|
Brady™
173384 |
2 x 1 in. | M710, BMP71 | 50.8 mm | Rectangle |
34.95 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
17858384
|
Brady™
174247 |
2.75 x 1.25 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 69.85 mm | Rectangle |
53.70 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||
Description
- Glossy white finish with permanent acrylic is thermal transfer printable
- UL approved, CSA accepted
- Meets requirements of a halogen-free material per DIN VDE 0472 part 815
- Service temperature of -70°C to 110°C (-94°F to 230°F)
- Made of B-423 polyester material
Specifications
Not Autoclavable | |
Permanent Acrylic | |
White | |
Die-cut | |
Single-side Printable | |
Labels | |
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory and Inspection Labeling | |
-94°F to +230°F | |
0.003 in. |
Halogen-free (DIN VDE 0472 Part 815) | |
None | |
Gloss | |
Polyester | |
Thermal Transfer | |
Non-sterile | |
No | |
-70°C to +110°C | |
0.076 mm |