Learn More
asecos™ Hazardous Substances Rack
The Hazardous Substances Rack provides storage of water pollutants with a steel sump on each storage level.
Brand:  asecos™ 27831
 View more versions of this product
Product Code. 15571445
Description
Description
  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • 4 painted sumps made of steel as storage levels
Specifications
Specifications
7.9 gal.
Steel
41.73 in.
41.73 in.
78.74 in.
25.08 in.
224.87 lb.
30 L
Drum Rack
1060.00 mm
1060.00 mm
2000.00 mm
637.00 mm
102 kg