missing translation for 'onlineSavingsMsg'
Learn More

Woodsafe Hazardous Waste Bin - Sharpsbin
GREENER_CHOICE

A cost-effective solution for lowering the CO2 emissions. Eco-friendly sharps and hazardous waste containers, produced from renewable resources using green energy. Reducing CO2 emissions by up to 66%, documented in a full LCA. Available in sizes from 0.5 L to 50 L.

Brand:  Woodsafe 1500120

 View more versions of this product

Product Code. 18772825

  • 2725.30 EUR / Pack of 96

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

Follow instructions for safe and correct use

This item is not returnable. View return policy

Description

Description

  • Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry.
  • Lowers the carbon footprint by more than 60%, proved in full LCA
  • Made from 100% Biocomposite
  • The lid can be temporarily closed to conceal the contents when not in use and securely permanently closed for transportation to the final disposal.
  • Natural colour and smell from wood - No migration of substances - EN - tested for toys and food
  • Compliant with UN3291
  • Tested according to ISO 23907-1:2019
  • UN ADR Approved
  • Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
  • NOTE: String between lids is for delivery purpose, remove/break before use!
  • Stop burning plastic
Specifications

Specifications

50 L
Temporary and permanent closure
Environmental friendly Sharpsbin
Brown, Yellow
Single-use
Yes
Non-sterile
Bin + Lid + Label
Pallet
Biohazard Waste Container (Benchtop)
EN ISO 23907-1:2019. UN3291, ADR Approved
Brown
13.2 gal.
39.5 x 39.5 x 49.5 cm (15.5 x 15.5 x 19.5 in.)
96
39.5 x 39.5 x 49.5 cm
Autoclavable
Safe Disposal of sharps and potentially Infectious Waste or medical waste
Biocomposite
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Voir les accessoires
Promotions

Promotions

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis

Lower climate impact