missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Woodsafe™ Sharpsbin and Hazardous waste containers with double lid in natural color

Woodsafe is a cost-effective solution for reducing the use of fossil based plastic by 80% and CO2 emissions by 66%, documented in in a full LCA. Produced with residue from the forest industry using green energy. Available in sizes from 0.5 L to 50 L. Single and double lid versions.
Brand: Woodsafe™ 1250120
Alert:
Follow instructions for safe and correct use
Description
- Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the Swedish forest industry
- Lowers the use of Fossil-based plastic by 80%!
- Lowers the carbon footprint by more than 60%, proved in full LCA
- Made from 100% Biocomposite
- The lid can be securely permanently closed for transportation to the final disposal
- My Green Labs ACT Label
- Follows same process as yellow bins and can be used to transport the same substances
- Natural color and smell from wood - No migration of substances
- Compliant with UN3291
- Tested according to ISO 23907-1:2019
- UN ADR Approved
- Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
- NOTE: String between lids is for delivery purpose, remove/break before use!
- Stop burning plastic
Specifications
25 L | |
Temporary and permanent closure | |
Environmental friendly Sharpsbin | |
Brown, Yellow | |
Single-use | |
Yes | |
Non-sterile | |
Bin + Lid + Label | |
Pallet | |
Biohazard Waste Container (Benchtop) |
EN ISO 23907-1:2019. UN3291, ADR Approved | |
Brown | |
6.6 gal. | |
39.5 x 39.5 x 23.8 cm (15.5 x 15.5 x 9.37 in.) | |
132 | |
39.5 x 39.5 x 23.8 cm | |
Autoclavable | |
Safe Disposal of sharps and potentially Infectious Waste or medical waste | |
Biocomposite |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lower climate impact and ACT label
Spot an opportunity for improvement?Share a Content Correction