Learn More
Lonza™ HBM™ Hepatocyte Basal Medium 500 mL
Unsupplemented basal medium designed to support the growth of human hepatocytes.
Brand:  Lonza™ CC-3197
Product Code. 11675450
  • 437.00 EUR / Each
Explore more special offers
Description
Description
  • Basal medium.
  • Contains no growth factors cytokines, or supplements.
  • Developed to support the growth of human hepatocytes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hepatocyte Cells
Basal Medium
500 mL