missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS, calcium, magnesium, phenol red

Product Code. p-200110074
Click to view available options
Quantity:
5 L
20 L
50 L
100 L
200 L
Format:
Drum
Sample
Tote
Unit Size:
100 L
20 L
200 L
50 L
Each
This item is not returnable. View return policy

Product Code. 30276209

Brand: Gibco™ A59772BA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order.

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order. Commonly used for processing cells and flushing equipment, sourcing Gibco balanced salt solutions can help streamline your operation by reducing buffer preparation and allowing you to focus on additional critical steps in your biopharmaceutical processes. Discover new outsourcing efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals
TRUSTED_SUSTAINABILITY

Specifications

Color Clear, colorless solution
Form Liquid
Product Type Hanks' Balanced Salt Solution (HBSS)
Sterility Sterile-filtered
Without Additives No Calcium, No Magnesium
Format Sample
Manufacturing Quality cGMP
Osmolality 265 to 295 mOsm/kg
pH 6.7 to 7.8
Quantity 5 L

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.