Learn More
Lonza™ HCM™ SingleQuots™ Kit
Supplements and growth factors kit for use with primary hepatocyte basal medium.
Brand: Lonza™ CC-4182
This item is not returnable.
View return policy
Description
- Separately sold SingleQuot™ kit for supplementing basal medium (CC-3199).
- Combined with basal medium, these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species.
- The final medium is serum-free
- Includes the following:
- Transferrin
- Ascorbic Acid Solution
- HEGF
- Recombinant Human Insulin (0.5%)
- Hydrocortisone
- BSA, Fatty Acid Free
- GA-1000
Specifications
Transferrin, Ascorbic Acid, HEGF, Recombinant Human Insulin (0.5%), Hydrocortisone, Fatty Acid-Free BSA, GA-1000 | |
Hepatocyte Cells | |
Supplements and growth factors |