missing translation for 'onlineSavingsMsg'
Learn More

Huenersdorf™ Polyethylene, Wide Corner Valve

Product Code. 10087600
Click to view available options
Length: 118mm
This item is not returnable. View return policy
This item is not returnable. View return policy

For Use With: Workshops, household and hobby

Order Info

dimLengthMetric

Specifications

Color Natural
For Use With (Equipment) Workshops, household and hobby
Length (Metric) 118 mm
Material HDPE
Material (Membrane) HDPE
No. per Pack 1 Ea.
Product Type Angle Valve
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'