Learn More
Brand™ HDPE Soaking Jar
Manufactured from High-Density Polyethylene. BRAND™ HDPE Soaking Jars are used with pipet rinsing systems and draining racks.
136.00 EUR - 151.00 EUR
Product Code | Brand | Capacity (Metric) | Height (Metric) | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Description | Price | Quantity & Availability | |||
10707791
|
Brand™
29210 |
10 L | 510 mm | For 460mm L pipets |
136.00 EUR
Each |
|
|||
10463901
|
Brand™
29215 |
12.5 L | 660 mm | For 600mm L pipets |
151.00 EUR
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
Description
- Manufactured from High-Density Polyethylene.
- Used with Pipet rinsing systems and draining racks.
Specifications
HDPE | |
240 mm Base |