missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE White Square Narrow Neck Bottle

Manufactured from HDPE for strength and durability. Kautex™ HDPE Square Narrow Mouth Bottles are easily stocked and save space in the laboratory from square shape.

Brand:  Kautex™ 2000083991

 View more versions of this product

Product Code. 15981441

  • 327.00 EUR / Pack of 122
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Square cross-section
  • Recessed labelling field
  • No cap. Four types of closures available separately
  • Ideal for liquids, leak-proof
Application:
  • packaging
  • sending samples
  • transport
  • storage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5 L
17 mm
HDPE
Narrow
170 mm
74 mm
White
Packaging, Sending Samples, Transport, Storage
Narrow
Square
Narrow Neck Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers