Learn More
Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations
Product Code. 11894770
Click to view available options
Holds:
4 x 96-well PTFE Microplates
40 x 24 mm Tubes
96 x 16 mm Tubes
missing translation for 'unitSize'
Each
Product Code. 11894770
missing translation for 'mfr': Cole-Parmer™ 9996805
Only null left
Add to basket
Add to basket
Accessory heating blocks to keep your reactions running smoothly.
  • Compatible with Electrothermal RS9000 Reaction Stations
One year warranty
  • Specifications
    Description Heating Block for Reaction System
    For Use With (Equipment) RS-250D-S Shaking Reaction Stations
    Holds 4 x 96-well PTFE Microplates
    Material PTFE
    Type Heating Block
    Includes 4 Blocks, 96 Wells
    Warranty 1 Year