missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEATR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5103883
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HEATR1.
Ribosome biogenesis factor. Involved in nucleolar processing of pre-18S ribosomal RNA. Required for optimal pre-ribosomal RNA transcription by RNA polymerase I. [UniProt]Specifications
HEATR1 | |
Polyclonal | |
Unconjugated | |
HEATR1 | |
AA517551; B130016L12Rik; BAP28; BC019693; FLJ10359; HEAT repeat containing 1; HEAT repeat-containing protein 1; HEAT repeat-containing protein 1, N-terminally processed; HEATR1; Protein BAP28; protein UTP10; RGD1564769; U3 small nucleolar RNA-associated protein 10 homolog; UTP10; UTP10, small subunit (SSU) processome component, homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
217995, 361262, 55127 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H583 | |
HEATR1 | |
A synthesized peptide derived from human HEATR1(Accession Q9H583), corresponding to amino acid residues E964-I1014. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |