Learn More
Hecht Karl™ Heavy-wall Test Tubes
Manufactured with AR-Glas™ (soda glass) to minimize breakage. Karl Hecht™ Heavy-Wall Test Tubes are suitable for general laboratory use.
Brand:  Hecht Karl™ 42775051
 View more versions of this product
Product Code. 10120071
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Soda lime glass
  • Round bottom
  • Rimless
  • Sized to support all laboratory needs 
Specifications
Specifications
Round Bottom
160 mm
100 Pack
15.25 mm
Test tube
Soda Glass
No