Learn More
Hecht Karl™ Heavy-wall Test Tubes
Manufactured with AR-Glas™ (soda glass) to minimize breakage. Karl Hecht™ Heavy-Wall Test Tubes are suitable for general laboratory use.
Brand:  Hecht Karl™ 42775024
 View more versions of this product
Product Code. 10292001
  • 18.00 EUR / Pack of 200
Explore more special offers
Description
Description
  • Soda lime glass
  • Round bottom
  • Rimless
  • Sized to support all laboratory needs 
Specifications
Specifications
Kahn
75 mm
200 Pack
12 mm
Kahn test tube
Soda Lime Glass
No