Learn More
Heidolph™ Hei-VAP Expert Control ML G3B XL
Adds numerous smart features to the Hei-VAP Expert, as well as an overview of all parameters and the current process.
Brand:  Heidolph™ 574-02315-00
Product Code. 17105364
  • 6555.00 EUR / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Description
  • 5 in. touch display for controlling the entire system including the vacuum and the cooling temperature
  • Motor lift model with transparent plastic coated G3 XL vertical glassware - achieve higher evaporation rates due to expanded cooling surface of 2,200 cm2
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
  • Direct access to two favorites on the start screen
  • Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
  • Define and save own evaporation processes (ramps)
  • Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
  • With USB and Micro SD interface for easy installation of software updates and transfer of individual favorites from one device to another
Specifications
Specifications
EU Plug
Motor Lift Rotary Evaporator