Learn More
Heidolph™ Instruments Hei-VAP Ultimate Control HL G1, EU-plug
The Hei-VAP Ultimate Control HL G1 adds numerous smart features to the Hei-VAP Ultimate.
Brand:  Heidolph™ Instruments 575-02100-00
Product Code. 17323350
  • 6095.00 EUR / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Description
Adds numerous smart features to the Hei-VAP Ultimate, as well as an overview of all parameters and the current process.
  • 7 in. touch display for controlling the entire system including the vacuum and the cooling temperature
  • Hand lift model with G1 diagonal glassware
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
  • Direct access to two favorites on the start screen
  • Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
  • Define and save own evaporation processes (ramps)
  • Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
  • USB and Micro SD plus LAN / RS 232 interfaces (Lab 4.0 ready)
Specifications
Specifications
5 L
Hand Lift Rotary Evaporator
EU-plug