Learn More
Motic™ Hemocytometer Holder
Firmly holds hemocytometer on your microscope stage. Motic™ Hemocytometer Holder facilitates hemocytometer viewing and systematic cell counting.
Brand:  Motic™ 1101000200292
Product Code. 15415332
  • 85.10 EUR / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Description
  • For use with AE Series Microscope stages.
  • Facilitates hemocytometer viewing and systematic cell counting.
Specifications
Specifications
Hemocytometer Holder
Hemocytometer (35 x 76 mm)
AE Series Microscope stages