Learn More
Motic™ Hemocytometer Holder
Firmly holds hemocytometer on your microscope stage. Motic™ Hemocytometer Holder facilitates hemocytometer viewing and systematic cell counting.
Brand: Motic™ 1101000200292
This item is not returnable.
View return policy
Description
- For use with AE Series Microscope stages.
- Facilitates hemocytometer viewing and systematic cell counting.
Specifications
Hemocytometer Holder | |
Hemocytometer (35 x 76 mm) | |
AE Series Microscope stages |