missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES hemisodium salt

Product Code. 15454329
Click to view available options
Quantity:
100 g
250 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A zwitterionic buffer

HEPES hemisodium salt is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES hemisodium salt is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103404-87-1
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Color White
Physical Form Crystalline Powder
Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 497.58
Chemical Name or Material HEPES hemisodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.