Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories
Shop All Thermo Scientific ProductsDescription
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Specifications
Specifications
| For Use With (Equipment) | Heraguard ECO 1.2 |
| Depth (Metric) | 580 mm |
| Description | Stainless Steel Work Surface |
| Width (Metric) | 1200 mm |
| Product Type | Work Surface Wraps |
| Configuration Options | Factory Installed |