missing translation for 'onlineSavingsMsg'
Learn More

Hexane, mixture of isomers, Puriss., ≥85% isomers basis (GC), Honeywell™

Product Code. 15661490 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15661490

Brand: Honeywell Research Chemicals 156131L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Puriss.,≥85% isomers basis (GC)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Density 0.672g/mL at 25°C
Quantity 1 L
Explosion Limit 0.075
Flash Point −23°C (−9.4°F)
Ignition Point 437°F
UN Number 1208
Beilstein 1730733
Formula Weight 86.18g/mol
Percent Purity ≥85% isomers basis (GC)
Grade puriss.
Chemical Name or Material Hexane, Mixture Of Isomers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.