missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Aluminum Bottle
Unit Size:
1 L
4 L
Specifications
Specifications
| Density | 0.663g/mL at 25°C (typical) |
| Boiling Point | 68.7°C |
| Quantity | 4 L |
| Flash Point | −23.00°C |
| Packaging | Aluminum Bottle |
| Specific Gravity | 1L = 0.66kg |
| Formula Weight | 86.18 |
| Grade | Meets ACS reagent chemical requirements |
| Chemical Name or Material | Hexanes |
Safety and Handling
Product Identifier
- n-hexane
- Danger
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H361f-Suspected of damaging fertility.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction