Learn More
WTW™ Hg-free Reaction Cuvette
144.00 EUR
Specifications
Type Hg-free Reaction Cuvette
Diameter (Metric) 16 mm
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10629363
View Documents
WTW™
250301
Measuring Range: 10 to 150 mg/L COD
144.00 EUR
Each
10677813
View Documents
WTW™
250306
Measuring Range: 100 to 1500 mg/L COD
144.00 EUR
Each
Specifications
Hg-free Reaction Cuvette
16 mm